Learn More
Lenz Laborglasintrumente Spiral Condensers, with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1020.00 SEK - 1620.00 SEK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11556804
|
Lenz Laborglasintrumente
|
160 mm |
1020.00 SEK
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||||
10624915
|
Lenz Laborglasintrumente
05233414 |
400 mm |
1555.00 SEK
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
11586804
|
Lenz Laborglasintrumente
05233514 |
500 mm |
1620.00 SEK
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||||
Description
- Spiral condensers for cooling condensates
- With NS socket and NS cone
- Water connection: GL 14 threads (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
GL 14 threads and two threaded plastic olives |