Learn More
Hettich Lab Technology™ Square Carrier
Square Carrier/Bucket is suitable for Swing-out Rotor 56999-R and accepts a range of adapters for tubes and plates of varying capacity.
Brand:  Hettich Lab Technology™ 5628
Product Code. 10069741
  • 5895.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Open bucket/carrier for adapter inserts
  • Suitable for several swing-out rotors and adapters
  • Accommodates a variety of centrifuge workloads
Specifications
Specifications
Swing-Out Rotor 5699-R, Adapter 5227, 5242, 5243, 5249, 5258, 5262, 5263-A, 5264, 5266, 5267, 5268, 5280, 5281, 6337-B, 6338-B, 6339-A, MTP, CP, DWP, MS, QP, Microtest plate (Terasaki), 96-well PCR Plate, PCR Strips
Bucket
Carrier, Bucket
1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hettich Lab Technology™ Square Carrier > No. of Places: 1