Learn More
Gilac™ Square Bucket with Watertight Lid, 12 L
Square 12 L Bucket with Watertight Lid fit for food contact with watertight lid and pouring spout.
370.93 SEK
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
17998511
|
Gilac™
G615712 |
Color: White |
370.93 SEK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17908861
|
Gilac™
G615718 |
Color: Yellow |
370.93 SEK
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17998431
|
Gilac™
G615710 |
Color: Blue |
370.93 SEK
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17978671
|
Gilac™
G615711 |
Color: Green |
370.93 SEK
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Bucket fit for food contact with watertight lid and pouring spout.
- Airtight lid keeps flavours in and odours out.
- Nesting when empty, no suction and stacking when lid is closed.
- Enables liquid preparations to be stored and transported.
- Graduation in litres.
- Discharge area that prevents water retention when being cleaned and anti-wear skirt that lifts the bucket off the ground.
- HDPE body and LDPE lid.
- Stainless steel handle.
- External dimensions include handle.
Specifications
Bucket, Handle, and Lid | |
12 L | |
Square |