missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 7826
Potent and selective LIMK inhibitor; antitumor
3040.00 SEK - 11925.00 SEK
Specifications
Chemical Name or Material | N-(2-Hydroxyethyl)-N′-[4,5-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-yl)phenyl]-N-phenylurea |
---|---|
CAS | 1219728-20-7 |
Target | LIMK Inhibitors |
Molecular Formula | C22H21N5O2 |
Formula Weight | 387.43 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784079
|
Tocris Bioscience™
5626/50 |
50 mg |
11925.00 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700498
|
Tocris Bioscience™
5626/10 |
10 mg |
3040.00 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective LIMK inhibitor (IC50 values are 43, 5536 and 6565 nM for LIMK1, ROCKI and ROCKII, respectively). Inhibits cofilin phosphorylation in A7r5 cells and suppresses migration and invasion of PC-3 cells in vitro. Orally bioavailable.Specifications
N-(2-Hydroxyethyl)-N′-[4,5-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-yl)phenyl]-N-phenylurea | |
LIMK Inhibitors | |
387.43 |
1219728-20-7 | |
C22H21N5O2 | |
>98% |
Safety and Handling
Recommended Storage : Store at 4°C