Learn More
Bochem™ Stainless Steel Beakers With Spout and Handle
Click to view available options
Stainless Steel Beaker; Capacity: 5000 mL
Description
- 18/10 stainless steel
- Special-contact base for hotplates and magnetic stirrers
- With pouring spout and handle
- 5000 mL capacity
Specifications
Specifications
Description | Stainless Steel Beaker |
Capacity (Metric) | 5000 mL |
Material | Stainless Steel (18/10) |
For Use With (Equipment) | General Lab Use, Hotplates and Magnetic Stirrers |
Includes | Rim, Pouring Spout and Handle |
Type | Beaker |