Learn More
Memmert™ Stainless Steel Bottom Drip Tray for UF160plus, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand: Memmert™ UF160plusB04359
This item is not returnable.
View return policy
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
UF160plus Oven | |
Bottom Drip Tray | |
Stainless Steel | |
Rim: 15 mm |
Product Content Correction
Product Title
Memmert™ Stainless Steel Bottom Drip Tray for UF160plus, 15 mm > For Use With: UF160plus
Spot an opportunity for improvement?Share a Content Correction