missing translation for 'onlineSavingsMsg'
Learn More

3M™ Stainless Steel Cleaner & Polish Aerosol

Stainless Steel Cleaner & Polish Aerosol

Brand:  3M™ 7000116805

Additional Details : Weight : 7.20000kg Transport : UN number : 1950 Chem class : 2.1


Product Code. 17237630

  • 2690.00 SEK / 12 canisters
Estimated Shipment: 23-07-2024
to see stock.

Description

Description

  • Polishes and protects all stainless steel surfaces in one step
  • The spray clings to vertical surfaces and leaves a light, non-greasy film that resist fingerprints and marking
  • Does not contain harmful chemicals and safe to use in food preparation areas
  • Water based with a pleasant citrus fragrance
Specifications

Specifications

Cleaner
600 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable and non-flammable aerosols category 1
  • Gases under pressure
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H222-Extremely flammable aerosol.
  • H229-Pressurised container: May burst if heated.
  • H280-Contains gas under pressure; may explode if heated.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers