missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Stainless Steel Evaporating Dish
320.00 SEK - 442.00 SEK
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
10142074
|
Bochem™
8395 |
55 mm |
320.00 SEK
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
10645305
|
Bochem™
8397 |
65 mm |
442.00 SEK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
10525124
|
Bochem™
8396 |
60 mm |
379.00 SEK
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Specifications
Rectangular | |
Evaporating Dish |