Learn More
Trajan Scientific and Medical™ Stainless-Steel GC Unions
Provide low dead volume connections for optimum chromatography
Brand: Trajan Scientific and Medical™ 103330
This item is not returnable.
View return policy
Description
- Low dead volume
- For high temperature applications
- Leak-free connections
- Does not require back ferrules
- Used with Graphite, Graphite Vespel™ or PTFE ferrules
- Suitable for glass, fused silica, metal, plastic or glass lined tubing
- Non-swaging and no tube distortion
Specifications
Union | |
0.0625 x 0.0625 in. | |
Stainless Steel |