Learn More
Cole-Parmer™ Stainless Steel Luer Fittings
Exceptionally durable Luer fittings suitable for harsh chemical environments. Cole-Parmer™ Stainless Steel Luer Fittings are easy to disassemble for cleaning.
Brand: Cole-Parmer™ 41507-34
This item is not returnable.
View return policy
Description
Exceptionally durable Luer fittings suitable for harsh chemical environments. Cole-Parmer™ Stainless Steel Luer Fittings are easy to disassemble for cleaning.
- Exceptional durability
- Easy to disassemble for cleaning
- Ideal for harsh chemical environments
- Autoclavable
Specifications
Luer Fitting | |
Stainless Steel | |
1 Pack |
Autoclavable | |
Harsh chemical environments | |
Female Luer x 1/4-28 UNF thread |