Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UF110plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ UF110plusB02073
Product Code. 17508965
  • 1580.00 SEK / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
UF110plus Oven
Drip Tray
Stainless Steel
Rim: 15 mm
Documents
One moment while we fetch your results.
Product Title
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UF110plus, 15 mm > For Use With: UF110plus