Learn More
asecos™ Stainless Steel Small Parts Cleaner
The Stainless Steel Small Parts Cleaner is ideal for cleaning or degreasing small parts.
Brand:  asecos™ 40449
 View more versions of this product
Product Code. 17856556
  • 2800.00 SEK / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Description
  • With recessed grip
  • Spring-loaded immersion sieve incl. flame arrester, grounding connection
Specifications
Specifications
Parts Cleaner
8 L
Stainless Steel
255.00 mm
264.00 mm
2.3 kg
2.1 gal.
Gray
10.04 in.
10.39 in.
5.07 lb.