Learn More
Stainless Steel Spheres, 4mm (0.16 in.) dia., Thermo Scientific Chemicals
282.30 SEK - 985.00 SEK
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Appearance | Spheres |
Molecular Formula | Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11345959
|
Thermo Scientific Alfa Aesar
042021.KM |
25 Pc. | N/A | ||||||
11355959
|
Thermo Scientific Alfa Aesar
042021.KN |
100 Pc. |
985.00 SEK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt% | |
Insoluble in water. |
Spheres | |
MFCD00148499 | |
Sphere |
Safety and Handling
H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures