Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Square Holes
Brand: RETSCH 60.190.050000
This item is not returnable.
View return policy
Specifications
1 Each | |
Test Sieve | |
Certificate indicating Compliance | |
For separation, fractioning, particle size determination | |
Stainless Steel |
50 mm | |
ISO 3310/2 | |
50mm | |
200 mm | |
Stainless Steel |
Product Content Correction
Product Title
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Square Holes > Pore Size: 50 mm
Spot an opportunity for improvement?Share a Content Correction