Learn More
Brannan™ Bimetal Dial Thermometer with Spiked Stem
Brannan™ Bimetal Dial Thermometer with Spiked Stem is constructed of stainless steel and is suitable for use with corrosive liquids.
Brand: Brannan™ 31/288/0
This item is not returnable.
View return policy
Description
- Bimetal dial test thermometer with spike stem
- Stainless steel stem suitable for corrosive liquids
- For handheld use and general test measurement
Specifications
51 mm | |
200 mm | |
Stainless Steel | |
40°C | |
-40°C to +40°C |
4 mm | |
Plastic | |
Thermometer | |
-40°C |