Learn More
Bochem™ Stainless Steel Tubing Clamps
Secure tubing before severing it using Bochem™ Stainless Steel Tubing Clamps in the laboratory.
Brand:  Bochem™ 2292
 View more versions of this product
Product Code. 10634101
  • 155.00 SEK / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Description
  • Self-tension
  • Ridges at the top
Specifications
Specifications
1 Ea.
Tubing Clamp
Stainless Steel
160 mm
Product Title
Bochem™ Stainless Steel Tubing Clamps > Length: 160mm