Learn More
Atrow™ Stainless Steel Wire Basket with Handle
Load materials for storage, cleaning, chemical baths and more. Atrow™ Stainless Steel Wire Basket with Handle is available in a cylindrical or square shape in a variety of sizes. Each is made of vertical stainless steel bars and features a hinged handle for easy handling.
Brand: Atrow™ 811441
Additional Details : Weight : 0.26000kg
Description
Stainless steel is durable, resistant to rust and corrosion, and can be used in a wide range of temperatures. This enables these material handling baskets to be used in a wide range of applications, including workspace organization, cleaning and rinsing, chemical baths and temperature cycling.
- Round baskets with diameters from 140mm–300mm
- Square baskets with side lengths of 125mm–200mm
- Baskets can be submerged and used in high and low temperature environments
Specifications
Handle | |
Stainless Steel | |
1 Each | |
140 mm |
Wire Basket | |
Round | |
140mmID x 140mmH | |
140 mm |