Learn More
Invitrogen™ eBioscience™ StainTray™
May be used as a humidified, light-protected chamber for staining slides for immunohistochemistry or immunocytochemistry. The StainTray™ has a black base made of tough ABS plastic withstanding a wide range of chemicals (avoid chlorinated hydrocarbons).
Brand: Invitrogen™ 44-0404-10
2261.00 SEK valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accepts up to 20 slides on four plastic rails covered with a polymer strip to perfectly hold slides even if the tray is held at an angle
- When humidity is needed, wells between rails will hold up to 1mL of water securely without splashing
- Middle wells will hold up to 2mL each
- Rails are raised not only to avoid water touching the slides but also to make slides more easily retrievable
- The base will also hold excess stain solution dripping from the slides
- Four rubber feet ensure greater base stability
- Units are stackable for storage and space saving purposes
- A black lid is included for work with fluorescent reagents. It is made of ABS and can toleratae a temperature range of −80°C to 80°C
Specifications
Stain Tray | |
eBioscience™ | |
1 each |
20 | |
Immunocytochemistry, Immunohistochemical Staining, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Microscopy |
Technical Data Sheet: StainTray™
Version: 2/9/2015
File size: 286.4 kB
Product Content Correction
Product Title
Invitrogen™ eBioscience™ StainTray™ > Staintray