Learn More
Standard Barium, 1000ppm, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Concentration | 1000 ppm |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
FINA22E1 | Certificate of Analysis | 30/01/2024 | FINA1 |
Product Content Correction
Product Title
Standard Barium, 1000ppm, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction