missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2 Mouse anti-Human, Clone: STAT2/2650, Novus Biologicals™
Mouse Monoclonal Antibody
2450.00 SEK
Specifications
Antigen | STAT2 |
---|---|
Clone | STAT2/2650 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
STAT2 Monoclonal specifically detects STAT2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
STAT2 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
6773 | |
IgG1 κ | |
Protein A or G purified |
STAT2/2650 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
Recombinant full-length human STAT2 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title