Learn More
Brady™ Static Dissipative Thermal Transfer Printable Labels
Printable, static dissipative polyester labels for Thermal Transfer Printers.
Brand:  Brady™ 029905
 View more versions of this product
Product Code. 11513664
  • 2385.00 SEK / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Material is UL and CSA approved with its respective ribbon
  • Ideal for Circuit Board Labeling
  • For smooth surfaces
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Gloss
Thermal Transfer
Circuit Board Labeling, Electrical Labeling, General Identification
16.8 mm
10,000 Labels
-40°C to +120°C
13.6 mm
12.7 mm
11.1 mm
THT-12-473-10
White
Static-dissipative Polyester
Non-sterile
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
5
Rectangle
0.09 mm
12.7 mm
11.1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Static Dissipative Thermal Transfer Printable Labels > Static Dissipative Thermal Transfer Printable Labels; Labels per Row: 5; Shape: Rectangle; Height (Metric): 11.1 mm; Vertical Repeat (Metric): 13.6 mm; Width (Metric): 12.7 mm