missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearic Acid Monoethanolamide, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
CAS | 111-57-9 |
Molecular Formula | C20H41NO2 |
Quantity | 100 g |
Synonym | n-2-hydroxyethyl octadecanamide, n-stearoylethanolamine, stearoylethanolamide, stearoyl ethanolamide, n-2-hydroxyethyl stearamide, stearamide mea, stearamyl, stearic acid monoethanolamide, stearic monoethanolamide, stearic ethanolamide |
InChI Key | OTGQIQQTPXJQRG-UHFFFAOYSA-N |
SMILES | CCCCCCCCCCCCCCCCCC(=O)NCCO |
IUPAC Name | N-(2-hydroxyethyl)octadecanamide |
Molecular Weight (g/mol) | 327.553 |
PubChem CID | 27902 |
ChEBI | CHEBI:85299 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'