Learn More
Bochem™ Steel Stand Bases
Steel Stand Bases with one adjustable foot.
Brand:  Bochem™ 5037
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10588272
  • 610.00 SEK / Each
Estimated Shipment: 25-09-2024
to see stock.
Add to basket
Description
Description
  • Thread M10
  • 1 foot adjustable
Specifications
Specifications
Steel
315 mm
Stand Base
8 mm
200 mm
1 Ea.