Learn More
Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes
Conduct applications with the chemical resistance of Pyrex™ borosilicate glass using Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes, in easy to handle shelf packs.
Description
Conduct applications with the chemical resistance of Pyrex™ borosilicate glass using Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes, in easy to handle shelf packs.
Features:- Calibrated for delivery with the last drop expelled by blowing
- Available either plugged or unplugged
- Easy to read graduations
Ordering Information:
Product is not available in all geographic regions.
Specifications
Glass | |
Yes | |
290 mm |