Learn More
Thermo Scientific™ Sterilin™ Silicone Tubing
Perform various applications with high quality, silicone tubing made from nontoxic material.
Brand:  Thermo Scientific™ TSR0200100P
 View more versions of this product
Product Code. 12882983
  • 1615.00 SEK / 15m
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Description
Perform various applications with high quality, silicone tubing made from nontoxic material.
  • Laser etched batch code for ease of traceability
  • Ideal for use with peristaltic pumps
  • Can be autoclaved or dry heat sterilized
  • 15m length
  • Manufactured in ISO Class 7 clean room
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Silicone tubing
2 mm
4 mm
15 m
Translucent
Flexible Tubing
Autoclavable
Silicone
49 ft.
1 mm
Very Flexible