Learn More
Sampling Systems™ SteriWare™ Powder Thief
Learn how to prevent cross contamination when sampling when you use a SteriWare™ PowderThief sampler. With a SteriWare™ PowderThief sampler, you can take samples from most free-flowing powders and granules.
Brand: Sampling Systems™ 8060H-1520S
Description
- The sterile, disposable PowderThief is ideal for taking representative samples from known locations within the bulk powder.
- These samplers are available in two convenient lengths and four sample volumes.
- Disposable Powder Thief samplers are available sterile or non-sterile.
- Assembled and packed in a cleanroom.
- Individually bagged.
- Designed for single use.
- Made from FDA and EU conforming HDPE.
- BSE/TSE Free.
- Full Batch Traceability.
- Available as gamma irradiated sterile (SAL 10-6).
Specifications
HDPE | |
Single-use | |
Individually Bagged |
20 mL | |
Sterile | |
1500 mm |