Learn More
Lenz Laborglasintrumente Stirrer with Stirrer Bearing
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06310505
Product Code. 10649523
  • 1176.60 SEK / Each
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Shaft with blade and PTFE security screw
  • Gastight closure of vacuum stirring apparatuses up to 10-3 mbar at a continuous operation speed of max.
  • 800 rpm, for round bottom flasks 250-2000 mL
  • Shaft made of DURAN™ tubing
Specifications
Specifications
Sealing
1 Ea.
PTFE, Graphite