Learn More
IBA Lifesciences Strep-Tactin™ protein as lyophilized powder
GREENER_CHOICE
Strep-Tactin™ protein as lyophilized powder can be used for reversible immobilization of biotinylated analytes or Strep-tag™II and Twin-Strep-tag™ fusion proteins
Brand:  IBA Lifesciences 2-1204-005
 View more versions of this product
Product Code. 17171921
  • 4120.00 SEK / 5mg
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Strep-Tactin™ is a streptavidin variant, which can be utilized for coating of different surfaces. It is optimized for the binding of Strep-tag™II and Twin-Strep-tag™ fusion proteins but still maintains the ability to capture biotinylated targets. Therefore, it can be used for assay development and protein interaction studies, including experiments such as proximity-dependent biotin identification (BioID).
Specifications
Specifications
>50 nmol/mg
Lyophilized