Learn More
Sampling Systems™ Sub Sampler Pro
Can sample from a set distance from the bottom of a container or alternatively at a mid point in a container or well. Sampling Systems™ Sub Sampler Pro is sometimes called a Bomb Sampler and it is used throughout the world for both chemical and environmental sampling.
Brand: Sampling Systems™ 5072A-500
This item is not returnable.
View return policy
Description
- Lower Sub Sampler Pro until the plunger assembly reaches the bottom of the container
- When the plunger assembly hits the bottom, it is pushed up allowing the sample to enter the device
- When the sampler is raised the plunger will drop down and seal the sampler
- Lower the Sub Sampler Pro into the liquid
- At the required depth pull on the control line to raise the plunger
- This will allow the liquid to enter the sampler
- Release the control Line and the plunger will drop down and seal the sample
Specifications
Sampler | |
Nickel-plated Brass | |
500 mL |