missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxazole, 100%, MP Biomedicals™
749.00 SEK - 2785.00 SEK
Chemical Identifiers
CAS | 723-46-6 |
---|---|
Molecular Formula | C10H11N3O3S |
Molecular Weight (g/mol) | 253.276 |
InChI Key | JLKIGFTWXXRPMT-UHFFFAOYSA-N |
Synonym | sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin |
PubChem CID | 5329 |
ChEBI | CHEBI:9332 |
IUPAC Name | 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide |
SMILES | CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435131
|
MP Biomedicals
0215671125 |
25 g |
749.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445131
|
MP Biomedicals
0215671180 |
100 g |
2785.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
723-46-6 | |
253.276 | |
sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin | |
CHEBI:9332 | |
CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N |
C10H11N3O3S | |
JLKIGFTWXXRPMT-UHFFFAOYSA-N | |
5329 | |
4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide |
Specifications
723-46-6 | |
C10H11N3O3S | |
JLKIGFTWXXRPMT-UHFFFAOYSA-N | |
4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide | |
5329 | |
253.3 | |
Sulfamethoxazole |
White | |
sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin | |
CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N | |
253.276 | |
CHEBI:9332 | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulfamethoxazole
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphamethoxazole
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title