missing translation for 'onlineSavingsMsg'
Learn More

Sulfisoxazole, 99.0 - 101.0 %, MP Biomedicals™

Product Code. 11483371
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 127-69-5
Molecular Formula C11H13N3O3S
Molecular Weight (g/mol) 267.303
InChI Key NHUHCSRWZMLRLA-UHFFFAOYSA-N
Synonym sulfisoxazole, sulfafurazole, sulphafurazole, sulfisoxazol, sulfafurazol, sulfaisoxazole, sulfofurazole, sulfisoxasole, sulphaisoxazole, sulfadimethylisoxazole
PubChem CID 5344
ChEBI CHEBI:102484
IUPAC Name 4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzenesulfonamide
SMILES CC1=C(ON=C1C)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Assay Percent Range 99.0 to 101.0 %
Quantity 100 g
Formula Weight 267.3
Physical Form Crystalline
Chemical Name or Material Sulfisoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'