Learn More
Sulforhodamine B sodium salt, Thermo Scientific Chemicals
Product Code. 11477836
Click to view available options
Quantity:
5 g
25 g
Fluorescent dye which uses laser-induced fluorescence for quantification of cellular proteins
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3520-42-1
Molecular Formula C29H33N2NaO7S2
Molecular Weight (g/mol) 608.70
MDL Number MFCD00010180
InChI Key XJENLUNLXRJLEZ-UHFFFAOYSA-M
Synonym C.I. 45100; Kiton Red S
PubChem CID 131852807
IUPAC Name 2-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]-5-sulfobenzenesulfonate;sodium
SMILES [Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C
Specifications
Quantity 25 g
Beilstein 3886008
Formula Weight 580.66
Chemical Name or Material Sulforhodamine B sodium salt