missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid 2.5 M (5.0 N) Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA

Fully factorized, high purity, stable, certified product, developed and tested for acid/base titrations.

259.00 SEK - 774.00 SEK

Specifications

Concentration 2.5 M (5.0 N)
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11523012
BIOPHARMA
View Documents
Reagecon
SU22501
1 L HDPE Bottle GL45
259.00 SEK
1 L
Please to purchase this item. Need a web account? Register with us today!
11513012
BIOPHARMA
View Documents
Reagecon
SU22505
5 L HDPE Jerrycan
774.00 SEK
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
  • Acid/base titrations, also called neutralization titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.
Specifications

Specifications

2.5 M (5.0 N)
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • sulphuric acid 5.0N (2.5M)

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid

SDS
Documents

Documents

Product Certifications