missing translation for 'onlineSavingsMsg'
Learn More

Sumanirole maleate, Tocris Bioscience™

Product Code. 18083972
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective D2 agonist

High affinity D2 receptor agonist (EC50 values are between 17 and 75 nM in cell-based assays). Originally reported to exhibit >200-fold selectivity for the D2 receptor versus other dopamine receptor subtypes (Ki values are 9.0, 1940, >2190 and >7140 for D2, D3, D4 and D1 receptors respectively). More recently reported to be around 32-fold selective for D2 over D3 receptors (Ki values are 17.1 and 546 nM respectively). Exhibits antiParkinsonian activity.

Chemical Identifiers

CAS 179386-44-8
Molecular Formula C15H17N3O5
Molecular Weight (g/mol) 319.317
InChI Key VOJRMYBBPKNLLI-ORHWHDKWSA-N
Synonym sumanirole maleate, unii-2oi87t3r1b, u 95666e sumanirole maleate, r-5,6-dihydro-5-methylamino-4h-imidazo 4,5,1-ij quinolin-2 1h-one z-2-butenedioate, u 95666e, sumanirole maleate, r-5,6-dihydro-5-methylamino-4h-imidazo 4,5,1-ij quinolin-2 1h-one 2z-2-butenedioate, maleate, sumanirole maleate hplc, 5,6-dihydro-5-methylamino-4h-imidazo 4,5,1-ij quinolin-2 1h-one, r-maleate
PubChem CID 9818478
SMILES CNC1CC2=C3C(=CC=C2)NC(=O)N3C1.C(=CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 319.31
Percent Purity >98%
Chemical Name or Material Sumanirole maleate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'