Learn More
Hettich Lab Technology™ Swing-out Rotor for ROTIXA 50 S Centrifuge, 2-place
Designed for use with ROTIXA 50 S Centrifuge. These rotors are ideal for high sample throughput in medical, research and quality control laboratories.
Brand: Hettich Lab Technology™ 4282
Additional Details : Weight : 0.01000kg
Description
- Swing-out Rotor, 2-place
- For Deep Well Plates
- For ROTIXA 50 S Centrifuge
- ROTIXA models are the centrifuges of choice for medical routine laboratories, as well as research and quality control laboratories in the life science industry.
- ROTIXA centrifuges feature the extended S control panel.
- Ideal for high sample throughput.
Specifications
Swing-out Rotor | |
2,652 x g | |
183 mm | |
ROTIXA 50 S Centrifuge | |
2 |
20 Plates/Racks per Rotor | |
3,600 rpm | |
178 mm | |
90° |