Learn More
Hettich™ Swing-out Rotor for ROTIXA 500 RS Centrifuge, 4-place
Designed for use with ROTIXA 500 RS Centrifuge. These rotors are ideal for high sample throughput in medical, research and quality control laboratories.
Brand:  Hettich™
Product Code. 10384972
  • 27900.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Swing-out rotor, 4-place
  • For use with ROTIXA 500 RS Centrifuge
Specifications
Specifications
Swing-out Rotor
4,500 rpm
40 mm
4
5,252 x g
115 mm
ROTIXA 500 RS Centrifuge