Learn More
KARTELL Sylicon Conical Stopper
The Sylicon Conical Stopper creates a seal in bottles and flasks.
146.00 SEK - 2755.00 SEK
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Material | Silicone |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Top | Price | Quantity & Availability | |||||
10514631
|
KARTELL
0380000 |
9 mm |
146.00 SEK
Pack of 10 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10767292
|
KARTELL
0380200 |
13 mm |
169.00 SEK
Pack of 10 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10737482
|
KARTELL
0380500 |
18 mm |
200.00 SEK
Pack of 10 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10727672
|
KARTELL
0380700 |
23 mm |
430.00 SEK
Pack of 10 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10164811
|
KARTELL
0380800 |
26 mm |
605.00 SEK
Pack of 10 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10246241
|
KARTELL
0381200 |
42 mm |
1365.00 SEK
Pack of 5 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10747672
|
KARTELL
0381300 |
45 mm |
1485.00 SEK
Pack of 5 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10134861
|
KARTELL
0381400 |
48 mm |
1965.00 SEK
Pack of 5 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
10318371
|
KARTELL
0381500 |
54 mm |
2090.00 SEK
Pack of 5 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
10681312
|
KARTELL
0381600 |
60 mm |
2755.00 SEK
Pack of 5 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
Description
- Autoclavable.
- SUITABLE FOR FOODSTUFF.
- MATERIAL: SYLICON
Specifications
Autoclavable | |
White | |
Silicone | |
Non-sterile |