missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ T 112

Estrogen-related receptor γ agonist; promotes maturation of hiPSC-derived cardiomyocytes

1945.00 SEK - 7460.00 SEK

Specifications

Chemical Name or Material 2-[2-(4-Bromophenyl)-1H-benzimidazol-6-yl]-2,3-dihydro-5-(4-morpholinyl)-1H-isoindol-1-one
CAS 2376876-87-6
Target Estrogen and Related Receptor Agonists
Molecular Formula C25H21BrN4O2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18757086
View Documents
Tocris Bioscience™
7690/10
10 mg
1945.00 SEK
10mg
Please to purchase this item. Need a web account? Register with us today!
18757276
View Documents
Tocris Bioscience™
7690/50
50 mg
7460.00 SEK
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

T 112 is an estrogen-related receptor gamma (ERRγ) agonist. Promotes maturation of hiPSC-derived cardiomyocytes. T 112 accelerates metabolic and sarcomere maturation in cardiomyocytes, as well as maturation of contractile and electrophysiological properties and ultrastructure organization, when used alone or in combination with other small molecules.
Specifications

Specifications

2-[2-(4-Bromophenyl)-1H-benzimidazol-6-yl]-2,3-dihydro-5-(4-morpholinyl)-1H-isoindol-1-one
2376876-87-6
Estrogen and Related Receptor Agonists
C25H21BrN4O2
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.