Learn More
Thermo Scientific™ T Phosphoramidite, standard grade, serum vial bottle
T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis.
105.66 SEK - 367.60 SEK
Specifications
For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
---|---|
Product Type | T CEP |
Content And Storage | Store in freezer (at or below -20°C). |
Nucleoside Type | Deoxythymidine (dT) |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18044070
|
Thermo Scientific™
27-1736-04 |
1 g |
105.66 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17957172
|
Thermo Scientific™
27-1736-05 |
5 g |
367.60 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups (except T and U) designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxythymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C40H49N4O8P
• Base: T
• Formula weight: 744.82 g/mol
• CAS number: 98796-51-1
• HPLC Purity: ≥98.0%
• 31P NMR Purity: ≥98.0%
Specifications
AKTA Synthesizers, Applied Biosystems Synthesizers | |
Store in freezer (at or below -20°C). | |
Room Temperature | |
Phosphoramidite | |
DNA |
T CEP | |
Deoxythymidine (dT) | |
98% | |
Standard | |
Powder |
For Research Use Only. Not for use in diagnostic procedures.