missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF13 Monoclonal Antibody (2B3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA520069
This item is not returnable.
View return policy
Description
Peptide Sequence: KRKRLFSKEL RCMMYGFGDD QNPYTESVDI LEDLVIEFIT EMTHKAMSIG RQGRVQVEDI VFLIRKDPRK FARVKDLLTM NEELKRARKA FDEANYGS
TAF13 Monoclonal antibody specifically detects TAF13 in Human samples. It is validated for ELISA, Western Blot.Specifications
TAF13 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q15543 | |
TAF13 | |
TAF13 (NP_005636.1, 27 a.a. ∽ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2B3 | |
Unconjugated | |
TAF13 | |
1810004N01Rik; 2010309N11Rik; AI847295; MGC22425; TAF(II)18; TAF13; TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 18kDa; TAF13 RNA polymerase II, TATA box-binding protein-associated factor; Taf2k; TAFII18; TAFII-18; TATA box binding protein (TBP)-associated factor, RNA polymerase II, K, 18kD; TATA box binding protein associated factor 13; TATA-box binding protein associated factor 13; TBP-associated factor, RNA polymerase II, K, 18kD; transcription initiation factor TFIID 18 kD subunit; transcription initiation factor TFIID 18 kDa subunit; Transcription initiation factor TFIID subunit 13 | |
Mouse | |
Affinity chromatography | |
RUO | |
6884 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |