Learn More
TAF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541502
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PSVQPIVKLV STATTAPPST APSGPGSVQK YIVVSLPPTG EGKGGPTSHP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%
TAF6 Polyclonal antibody specifically detects TAF6 in Human samples. It is validated for ChIP Assay, Western Blot.Specifications
TAF6 | |
Polyclonal | |
Unconjugated | |
TAF6 | |
80kDa; AW549759; DKFZp781E21155; MGC:8964; p80; RNA polymerase II TBP-associated factor subunit E; RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAF(II)70; TAF(II)80; TAF2E; Taf6; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAFII70; TAFII-70; TAFII80; TAFII-80; TAFII85; TATA box binding protein (Tbp)-associated factor, RNA polymerase II, E; TATA box binding protein (TBP)-associated factor, RNA polymerase II, E, 70/85kD; TATA box binding protein associated factor 6; TATA-box binding protein associated factor 6; TBP-associated factor 6; TBP-associated factor 6 isoform alpha; TFIID subunit p80; transcription initiation factor TFIID 70 kDa subunit; Transcription initiation factor TFIID 80 kDa subunit; transcription initiation factor TFIID subunit 6; wu:fb83e08; wu:fj30b0 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6878 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P49848 | |
TAF6 | |
Synthetic peptide directed towards the C-terminal of human TAF6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |