missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAFA4 Monoclonal Antibody (480103), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524195
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 2, 3, or 5 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
TAFA4 Monoclonal antibody specifically detects TAFA4 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
TAFA4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96LR4 | |
FAM19A4 | |
E. coli-derived recombinant human TAFA4 Ser35-Arg140 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
480103 | |
Unconjugated | |
FAM19A4 | |
C130034I18Rik; Chemokine-like protein TAFA-4; chemokine-like protein TAFA-4; protein FAM19A4; FAM19A4; Fam19a4 Tafa-4; family with sequence similarity 19 (chemokine (C-C motif)-like), member A4; family with sequence similarity 19 member A4, C-C motif chemokine like; family with sequence similarity 19, member A4; protein FAM19A4; TAFA chemokine like family member 4; TAFA4; Tafa-4; TAFA4 protein | |
Mouse | |
Protein A/G | |
RUO | |
151647 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |