missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TALDO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565496
This item is not returnable.
View return policy
Description
Immunogen sequence: FVGRILDWHVA NTDKKSYEPL EDPGVKSVTK IYNYYKKFSY KTIVMGASFR NTGEIKALAG CDFLTISPKL LGEL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.Specifications
TALDO1 | |
Polyclonal | |
Unconjugated | |
TALDO1 | |
dihydroxyacetone transferase; glycerone transferase; TAL; TALDO; TALDO1; TALDOR; TALH; TAL-H; testicular secretory protein Li 56; transaldolase; transaldolase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P37837 | |
TALDO1 | |
Recombinant Human TALDO1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only