Learn More
BRAND™ Tall Form Graduated Beaker with Spout
4809.81 SEK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Use promo code "23264" to get your promotional price.
Click to view available options
Capacity: 2000 mL
Description
- Constructed of borosilicate glass
- With graduation and spout
Specifications
Capacity (Metric) | 2000 mL |
Material | Borosilicate Glass |
Includes | Spout |
Type | Tall Form Beaker |
Autoclavable | Autoclavable |
Graduated | Yes |
Certifications/Compliance | DIN EN 285 |