Learn More
Thermo Scientific™ Tango™ H4-bla U2OS
The Tango™ H4-bla U2OS cells contain the human Histamine Receptor 4 (H4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Marca: Thermo Scientific™ K1465
Descrizione
The Tango™ H4-bla U2OS cells contain the human Histamine Receptor 4 (H4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ H4-bla U2OS cells are functionally validated for Z' and EC50 concentrations of 4-Methylhistamine. In addition, Tango™ H4-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifica
NP_067637.2 | |
Division-Arrested Cells | |
Tail from AVPR2 GPCR | |
Fluorescence | |
HRH4 | |
Single Cell Clone Origin | |
8 x 108 Cells | |
BLA (Beta-Lactamase) | |
HRH4, H4 | |
U2OS | |
GPCR |
Cell-based beta-arrestin recruitment | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ H4-bla U2OS cells • Tango™ H4-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ H4-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Tango™ H4-bla U2OS, 8 x 106 Cells, ∽8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage | |
FRET | |
Tango™ H4-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ H4-bla U2OS cells, Tango™ H4-bla U2OS assay protocol, certificate of analysis | |
GeneBLAzer™, Tango™ | |
End Point | |
U2OS | |
U2OS Cell | |
Tango™ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only. Not for use in diagnostic procedures.