Learn More
Tanshinone IIA, MedChemExpress
MedChemExpress Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Brand: Medchem Express HY-N0135
This item is not returnable.
View return policy
Description
Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.Specifications
Tanshinone IIA | |
Red | |
C19H18O3 | |
Dan Shen ketone | |
O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC=C4C)=O | |
294.34 | |
Research | |
COVID-19-immunoregulation |
568-72-9 | |
Research | |
50 mg | |
DMSO : 100 mg/mL (339.74 mM; Need ultrasonic) | |
294.34 | |
99.17% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.