missing translation for 'onlineSavingsMsg'
Learn More

Taurocholic Acid Sodium Salt, MP Biomedicals™

Product Code. 17811421
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for protein solubilization. Taurocholic Acid Sodium Salt, MP Biomedicals is an anionic detergent that is the conjugation product of cholic acid with taurine. It is the principal constituent of the bile of carnivorous animals.

Biochemical actions:

  • Non-sulfated bile salt
  • Physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp)
  • Na(+)/taurocholate cotransporter (NTCP)
  • MRP3 into the hepatic carnalicular
TRUSTED_SUSTAINABILITY

Specifications

CAS 145-42-6
Physical Form Powder
Melting Point 230°C (decomposition)(literature)
pH 6 at 7.5 (1% aq. soln.)
Quantity 500 g
Molecular Formula C26H44NNaO7S
Synonym sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
Solubility Information Soluble in water (100mg/mL).
InChI Key JAJWGJBVLPIOOH-VXFFEJGCSA-M
SMILES CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
Molecular Weight (g/mol) 537.688
PubChem CID 131632374
Formula Weight 537.7
Percent Purity ≥97%
Grade Practical
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 3-11mM
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'