missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX21/T-bet Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13700-1-AP-20UL
This item is not returnable.
View return policy
Description
T-bet, also known as TBX21, is a T-cell specific transcription factor containing a single T-box DNA-binding domain. It controls the expression of the TH1 cytokine, interferon-gamma. Both Th1 and Th17 cells are crucial in immune regulation and autoimmune disease development. T-bet initiates Th1 lineage development from naive Th precursor cells and directs T cell differentiation to Th1 versus Th17. T-bet is cooperating with Stat4 (signal transducer and activator of transcription 4) in programming chromatin architecture for Th1 gene expression. Genetic variations in T-bet are associated with susceptibility to aspirin-induced asthma and nasal polyps.Specifications
TBX21/T-bet | |
Polyclonal | |
Unconjugated | |
TBX21 | |
T bet, T box 21, T box protein 21, T PET, TBET, TBLYM, TBX21, TBX21,T bet, Transcription factor TBLYM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
30009, 57765 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JKD8, Q9UL17 | |
Tbx21 | |
TBX21,T-bet Fusion Protein Ag4618 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TBX21/T-bet Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction