missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 Monoclonal Antibody (4E215), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA191539
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The T Complex Polypeptide 1 (TCP-1) is approximately 60 kDa protein constitutively expressed in almost all eukaryotic cells, and is upregulated during spermatogenesis. It is found in the cytosol as a subunit of a hetero-oligomeric chaperone that is known to be involved in the folding of actin and tubulin. The family of proteins termed chaperonins act to recognize and stabilize polypeptide intermediates during folding, assembly and disassembly, and share many characteristics with Heat Shock Protein 70 (HSP70) including high abundance, induction by environmental stress, and ATPase activity. The chaperonin family includes the mitochondrial HSP60, Escherichia coli GroEL, the plastid Rubisco-subunit binding protein, and the archaebacterial protein TF55. The TCP-1 sequence shows nearly 40% identity to TF55, but only minimal similarity to HSP60 and GroEL.Specifications
TCP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% Sodium azide; pH 7.2 | |
P11983, P18279, P28480, Q32L40 | |
TCP1 | |
Rats were immunized with a purified recombinant mouse TCP1 alpha construct encoding the C-terminal half of the 1.8 kb full-length Tcp-1b gene expressed in E. coli. | |
100 μg | |
Primary | |
Bovine, Canine, Hamster, Mouse, Ovine, Rabbit, Rat | |
Antibody | |
IgG2c |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
'4E215 | |
Unconjugated | |
TCP1 | |
65 kDa antigen; AI528772; c-cpn; CCT; Cct1; Ccta; CCTalpha; CCT-alpha; chaperonin-containing T-complex alpha subunit TCP1; chaperonin-containing T-complex polypeptide alpha subunit; cytosolic chaperonin containing t-complex polypeptide 1; D6S230E; hypothetical protein; p63; tailless complex polypeptide 1; tailless complex polypeptide 1A; Tailless complex polypeptide 1B; t-complex 1; t-complex polypeptide 1; t-complex protein 1; T-complex protein 1 alpha subunit-like protein; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; T-complex protein 1, alpha subunit; Tcp1; Tcp-1; TCP-1-A; TCP-1-alpha; TCP-1-B; Tp63; TRic; Unknown (protein for MGC:133746); YD8142.13; YD8142B.04; YDR212W | |
Rat | |
Protein G | |
RUO | |
100135437, 100302435, 100689476, 21454, 24818, 484064, 512043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |