missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Buffer Solution pH 2, WTW™
Click to view available options
Quantity:
1 L
Packaging:
Bottle
Description
- pH buffer by WTW that is precise and traceable to PTB/NIST in bulk size of 1 liter with built-in dosing vessel standard buffer
- Live up to 3 years (unopened)
- Thrifty use
- Easiest handling by means of dosing device
- This special buffer pH 2.00 is available in 1 l bulk
- All modern technical buffering solutions are traceable to PTB (Physical Technical Government Agency) or NIST (National Institute of Standards and Technology, USA)
- Their accuracy is ± 0,03 pH
Specifications
Specifications
pH | 2 |
Physical Form | Liquid |
Quantity | 1 L |
Packaging | Bottle |
Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction